missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Tamper-Evident Fire Extinguisher Seal

Catalog No. 19103630
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19103630 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19103630 Supplier Brady™ Supplier No. 103630

missing translation for 'validMainframeMsg'

Deter pranksters from tampering. The Brady™ Tamper-Evident Fire Extinguisher Seal indicates tampering at a glance, yet easily breaks away upon activation.

Specifications

Product Type Tamper-proof Seal
Color Natural
Material Polypropylene
For Use With (Equipment) Fire-Extinguishers
Length (English) 5.5 in.
Quantity 100 Pk.