missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen, MP Biomedicals

Catalog No. p-7112294
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215673891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215673891 Supplier MP Biomedicals Inc Supplier No. 0215673891

Tamoxifen, White Powder

  • Tamoxifen has been used to facilitate the recombination of ect2flox allele in mouse organs
  • It has also been used to study its effect on lipopolysaccharide (LPS)-induced microglial activation

Chemical Identifiers

CAS 10540-29-1
Molecular Formula C26H29NO
Molecular Weight (g/mol) 371.52
MDL Number MFCD00010454
InChI Key NKANXQFJJICGDU-QPLCGJKRSA-N
PubChem CID 2733526
ChEBI CHEBI:41774
SMILES CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

Specifications

Melting Point 96°C to 100°C
Color White
Linear Formula C26H29NO
Quantity 1 g
Synonym tamoxifen, trans-tamoxifen, crisafeno, soltamox, tamoxifene, diemon, tamoxifeno, tamoxifenum, citofen, istubol
Solubility Information Practically insoluble in water (< 0.01% at 20°C); soluble in methanol, ethanol, 2-propanol, propylene glycol, chloroform (50mg/mL - clear, colorless to faint yellow solution) or DMSO.
IUPAC Name 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine
Formula Weight 371.5
Physical Form Powder
Chemical Name or Material TAMOXIFEN
Recommended Storage Store at +4°C.