missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tamoxifen Citrate 98.0+%, TCI America™
Supplier: TCI America T25105G
Specifications
| Tamoxifen Citrate | |
| 142°C | |
| C32H37NO8 | |
| 5 g | |
| FQZYTYWMLGAPFJ-OQKDUQJOSA-N | |
| (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid | |
| 2733525 | |
| 563.65 | |
| Crystalline Powder |
| 54965-24-1 | |
| White | |
| MFCD00058321 | |
| tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 | |
| 563.65 | |
| CHEBI:9397 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 54965-24-1 | |
| 563.65 | |
| FQZYTYWMLGAPFJ-OQKDUQJOSA-N | |
| 2733525 | |
| (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid |
| C32H37NO8 | |
| MFCD00058321 | |
| tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen | |
| CHEBI:9397 | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 |
Safety and Handling
RTECSNumber : KH2387000
TSCA : No
Recommended Storage : Refrigerator