missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen Citrate 98.0+%, TCI America™

Catalog No. p-7137251
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T25101G 1 g
T25105G 5 g
2 options

Catalog No. T25101G

Supplier: TCI America T25101G

Only null left

Chemical Identifiers

CAS 54965-24-1
Molecular Formula C32H37NO8
Molecular Weight (g/mol) 563.65
MDL Number MFCD00058321
InChI Key FQZYTYWMLGAPFJ-OQKDUQJOSA-N
Synonym tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen
PubChem CID 2733525
ChEBI CHEBI:9397
IUPAC Name (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

Specifications

Melting Point 142°C
Color White
Quantity 1 g
Formula Weight 563.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tamoxifen Citrate
RTECSNumber KH2387000
TSCA No
Recommended Storage Refrigerator