missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012378
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012378 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012378 Supplier Merck Emd Millipore Supplier No. 8525650MG
Only null left
Pharmaceutical (small molecule)

Specifications

CAS 10540-29-1
Physical Form Neat
Linear Formula C6H5 C(C2H5)=C(C6H5)C6H4 OCH2 CH2 N(CH3)2
MDL Number MFCD00010454
Melting Point 97°C to 98°C
Synonym (Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene; trans-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethylamine
Type Analytical Standard
Quantity 50 mg
Formula Weight 371.51
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less