missing translation for 'onlineSavingsMsg'
Learn More

Cytiva TALON™ Superflow Medium

Catalog No. 45002389
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002389 10 mL
45002390 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45002389 Supplier Cytiva Supplier No. 28957499

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offers different selectivity compared to nickel-charged media. Cytiva TALON™ Superflow Medium used for purification of histidine-tagged recombinant proteins by IMAC.

Features:

  • Ligand - TALON Superflow Cobalt-IMAC
  • Matrix - Cross-linked agarose, 6%
  • Particle Size - 60 -160μm
  • Binding Capacity/mL - Up to 20 mg histidine-tagged protein
  • pH stability working range - 3 to 12
  • pH stability Cleaning-in-Place (CIP) - 2 - 14 Co2+stripped medium
  • Complete packsize - 10mL

 

Specifications

Quantity 10 mL
For Use With (Application) For PurifiCatIon of Histidine-Tagged Recombinant Pby Immobilized Metal Affinity Chromatography
Storage Requirements 4°C to 8°C
Binding Capacity Up to 20 mg Histidine-tagged Protein
Product Type Talon Superflow Media
Particle Size 60 to 160 μm
Matrix 6% Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Talon Superflow Cobalt-IMAC