missing translation for 'onlineSavingsMsg'
Learn More

Talniflumate 98.0+%, TCI America™

Catalog No. T3101200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3101200MG 200 mg
T31011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3101200MG Supplier TCI America Supplier No. T3101200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66898-62-2
Molecular Formula C21H13F3N2O4
Molecular Weight (g/mol) 414.34
MDL Number MFCD00866135
InChI Key ANMLJLFWUCQGKZ-UHFFFAOYNA-N
Synonym talniflumate, somalgen, lomucin, talniflumato, talniflumate usan:inn, talniflumatum inn-latin, talniflumato inn-spanish, phthalidyl 2-3-trifluoromethylanilino nicotinate, 3-phthalidyl ester of 2-3-trifluoromethyl phenyl amino-3-pyridinecarboxylic acid, phthalidyl 2-alpha,alpha,alpha-trifluoro-m-toluidino nicotinate
PubChem CID 48229
IUPAC Name 3-oxo-1,3-dihydro-2-benzofuran-1-yl 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylate
SMILES FC(F)(F)C1=CC=CC(NC2=NC=CC=C2C(=O)OC2OC(=O)C3=CC=CC=C23)=C1

Specifications

Melting Point 168°C
Color Yellow
Quantity 200 mg
Formula Weight 414.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Talniflumate
RTECSNumber QT2999200
TSCA No
Recommended Storage Refrigerator