accessibility menu, dialog, popup

UserName

CAS RN 66898-62-2

IUPAC Name: 3-oxo-1,3-dihydro-2-benzofuran-1-yl 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylate
Synonyms: talniflumate talniflumato somalgen lomucin talniflumatum
Molecular Weight (g/mol): 414.34
Molecular Formula: C21H13F3N2O4
InChi Key: ANMLJLFWUCQGKZ-UHFFFAOYNA-N
SMILES: FC(F)(F)C1=CC=CC(NC2=NC=CC=C2C(=O)OC2OC(=O)C3=CC=CC=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results