Learn More
Invitrogen™ Tali™ Cellular Analysis Slides
The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer.
$407.00
Specifications
| Type | Cellular Analysis Slide |
|---|---|
| Product Line | Tali |
| For Use With (Equipment) | Tali™ Image-Based Cytometer |
| Detection Method | Fluorescence |
| Shipping Condition | Room Temperature |
Description
The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer. Each slide contains two separate, enclosed analysis chambers designed to accommodate 25 μL of sample per chamber. Each box contains 50 slides.
Ready to Use for Your Convenience
Each Tali™ Cellular Analysis slide is pre-sterilized and individually packaged. Since the Tali™ Image-Based Cytometer is a fluorescent cellular analysis instrument the slides are composed of low fluorescent plastic. The slides are disposable; after use the slides can be disposed of with you regular biohazard waste.
Order Info
Shipping Condition: Room temperature
Specifications
| Cellular Analysis Slide | |
| Tali™ Image-Based Cytometer | |
| Room Temperature |
| Tali | |
| Fluorescence |
For Research Use Only. Not for use in diagnostic procedures.