missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tali™ Cellular Analysis Slides

Catalog No. T10794
Encompass
Change view
Click to view available options
Quantity:
50 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10794 50 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10794 Supplier Invitrogen™ Supplier No. T10794

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer.

The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer. Each slide contains two separate, enclosed analysis chambers designed to accommodate 25 μL of sample per chamber. Each box contains 50 slides.

Ready to Use for Your Convenience
Each Tali™ Cellular Analysis slide is pre-sterilized and individually packaged. Since the Tali™ Image-Based Cytometer is a fluorescent cellular analysis instrument the slides are composed of low fluorescent plastic. The slides are disposable; after use the slides can be disposed of with you regular biohazard waste.

Order Info

Shipping Condition: Room temperature

Specifications

Type Cellular Analysis Slide
Product Line Tali
For Use With (Equipment) Tali™ Image-Based Cytometer
Detection Method Fluorescence
Content And Storage Contains one box of 50 slides. Store at room temperature.
Quantity 50 Slides
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.