Learn More
Agilent BioTek Take3 Trio Microvolume Plate
Description
Microvolume quantification is fast and easy by measuring multiple 2 μL samples at the same time, without diluting and without the need for specialized equipment. Preprogrammed nucleic acid and protein protocols in the Take3 app bring confidence in your quantification data.
- Low volume nucleic acid and protein quantification
- It takes only 2 μL of undiluted sample to get rapid results with the Take3 Trio plate. Take3 Trio plates are compatible with most Agilent BioTek multimode readers, imaging multimode readers, and microplate spectrophotometers.
- Easy to clean and add more samples
- Take3 Trio plates are so easy to clean – simply wipe samples off after measurement with a laboratory wipe.
- Read 2 μL microspots or BioCells
- Along with 48 microspots, the Take3 Trio plate can accept 2 patented BioCells for rapid 1 cm measurements in an Agilent BioTek microplate spectrophotometer.
- Take3 App
- Preprogrammed protocols in the Take3 app for dsDNA, ssDNA, RNA, and protein (BSA, IgG, lysozyme) measurement make getting direct quantitative results quick and easy. Rapid visual results are output in convenient tab delimited .txt formats.
- Instrument compatibility: Cytation C10, Cytation 7, Cytation 5, Cytation 1, Synergy Neo2, Synergy H1, Synergy HTX, Synergy LX, Epoch 2, Epoch
Specifications
Specifications
| Product Type | Microvolume Plate |
| For Use With (Equipment) | Microplate Reader |
| Plate Type | 48 x 2&muL; microspots and 2 BioCells |