missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAFA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149842
Description
TAFA4 Polyclonal Antibody for Western Blot, IHC, ICC/IF
This gene is a member of the TAFA family which is composed of five highly homologous genes that encode small secreted proteins. These proteins contain conserved cysteine residues at fixed positions, and are distantly related to MIP-1alpha, a member of the CC-chemokine family. The TAFA proteins are predominantly expressed in specific regions of the brain, and are postulated to function as brain-specific chemokines or neurokines, that act as regulators of immune and nervous cells. Alternatively spliced transcript variants have been observed for this gene.Specifications
| TAFA4 | |
| Polyclonal | |
| Unconjugated | |
| FAM19A4 | |
| C130034I18Rik; Chemokine-like protein TAFA-4; chemokine-like protein TAFA-4; protein FAM19A4; FAM19A4; Fam19a4 Tafa-4; family with sequence similarity 19 (chemokine (C-C motif)-like), member A4; family with sequence similarity 19 member A4, C-C motif chemokine like; family with sequence similarity 19, member A4; protein FAM19A4; TAFA chemokine like family member 4; TAFA4; Tafa-4; TAFA4 protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 151647 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96LR4 | |
| FAM19A4 | |
| Recombinant Family With Sequence Similarity 19, Member A4 (FAM19A4) (His46-Thr139). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |