missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAFA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149946
Description
TAFA3 Polyclonal Antibody for Western Blot, IHC, ICC/IF
The TAFA3 gene encodes a protein that is part of the TAFA family of chemokine-like proteins, involved in modulating immune responses and inflammatory processes. TAFA3 is particularly implicated in the regulation of neuroinflammatory and neuroprotective activities in the central nervous system. It functions as a signaling molecule that can influence cell survival, proliferation, and migration, contributing to homeostasis in neural tissue and promoting recovery following injury. TAFA3 has been studied for its role in neuroprotection, where it helps shield neurons from damage caused by oxidative stress and inflammatory stimuli, making it significant in neurodegenerative conditions. Apart from its neuroprotective capabilities, TAFA3 may also be involved in the systemic modulation of inflammation, affecting immune cell trafficking and responses. Research into TAFA3 continues to unveil its multifaceted role in health and disease, highlighting potential therapeutic avenues for targeting this protein in conditions characterized by excessive inflammation or neurodegeneration.Specifications
| TAFA3 | |
| Polyclonal | |
| Unconjugated | |
| FAM19A3 | |
| Chemokine-like protein TAFA-3; FAM19A3; family with sequence similarity 19 (chemokine (C-C motif)-like), member A3; family with sequence similarity 19 member A3, C-C motif chemokine like; protein FAM19A3; TAFA3; TAFA-3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 329731 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q7TPG6 | |
| FAM19A3 | |
| Recombinant Family With Sequence Similarity 19, Member A3 (FAM19A3) (Ser30-Lys129). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody |