missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAF15 Monoclonal Antibody (8TA-2B10)
GREENER_CHOICE

Catalog No. PIMA3078
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA3078 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA3078 Supplier Invitrogen™ Supplier No. MA3078
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

MA3-078 detects TATA-Binding Protein (TBP) Associated Factor 15 from human, mouse, rat, canine, and non-human primate samples. The MA3-078 immunogen is the RNA- binding region (RNP) of human TAFII68 as a GST-fusion protein (from aa 175 to 414).

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals.

Specifications

Antigen TAF15
Applications Flow Cytometry, Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 8TA-2B10
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene TAF15
Gene Accession No. Q92804
Gene Alias 2610111C21Rik; 68 kDa TATA-binding protein-associated factor; 68kDa; hTAFII68; LOC100713096; Npl3; RBP56; RBP56/CSMF fusion; RNA-binding protein 56; TAF(II)68; Taf15; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68 kDa; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68kDa; TAF2N; TAFII68; TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA binding protein 56); TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA-binding protein 56); TATA box binding protein associated factor 15; TATA box-binding protein-associated factor 2N (RNA-binding protein 56); TATA-bin; TATA-binding protein-associated factor 2N; TATA-box binding protein associated factor 15; TBP-associated factor 15
Gene Symbols TAF15
Host Species Mouse
Immunogen RNA binding region (RNP) of human TAFII68 as a GST-fusion protein (from amino acid 175 to 414).
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287571, 491135, 70439, 8148
Target Species Canine, Human, Mouse, Monkey, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less