missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tadalafil, 97%

Catalog No. AC461940050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461940050 5 g
AC461940010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461940050 Supplier Thermo Scientific Chemicals Supplier No. 461940050
Only null left

CAS: 171596-29-5 | C22H19N3O4 | 389.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 171596-29-5
Molecular Formula C22H19N3O4
Molecular Weight (g/mol) 389.41
InChI Key WOXKDUGGOYFFRN-IIBYNOLFSA-N
IUPAC Name (2R,8R)-2-(2H-1,3-benzodioxol-5-yl)-6-methyl-3,6,17-triazatetracyclo[8.7.0.0³,⁸.0¹¹,¹⁶]heptadeca-1(10),11,13,15-tetraene-4,7-dione
SMILES CN1CC(=O)N2[C@H](CC3=C(NC4=CC=CC=C34)[C@H]2C2=CC=C3OCOC3=C2)C1=O

Specifications

Melting Point 296.0°C to 303.0°C
Assay Percent Range 97%
Quantity 5 g
Solubility Information Solubility aq. soln.: <1 mg/mL Other solubilities: 78 mg/mL dmso, <1 mg/mL ethanol
Formula Weight 389.4
Chemical Name or Material Tadalafil

RUO – Research Use Only