accessibility menu, dialog, popup

UserName

CAS RN 171596-29-5

CH3NON(R)HN(R)OOO

CAS RN 171596-29-5

IUPAC Name: (2R,8R)-2-(2H-1,3-benzodioxol-5-yl)-6-methyl-3,6,17-triazatetracyclo[8.7.0.0³,⁸.0¹¹,¹⁶]heptadeca-1(10),11,13,15-tetraene-4,7-dione
Synonyms: tadalafil CIA cialis tadanafil cialis/taladafil
Molecular Weight (g/mol): 389.41
Molecular Formula: C22H19N3O4
InChi Key: WOXKDUGGOYFFRN-IIBYNOLFSA-N
SMILES: CN1CC(=O)N2[C@H](CC3=C(NC4=CC=CC=C34)[C@H]2C2=CC=C3OCOC3=C2)C1=O
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results