Learn More
Sartorius Tacta™ Mechanical Pipettes
Catalog No. p-6684012
Change view
Click to view available options
Number of Channels:
1
12
8
Volume (Metric):
0.1 to 3 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
1000 to 10,000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
5 to 100 μL
500 to 5000 μL
Color:
Blue
Gray
Green
Orange
Red
Yellow
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels Volume (Metric) Color
14577793 1 0.1 to 3 μL Gray
14577794 1 0.5 to 10 μL Gray
14577795 1 2 to 20 μL Yellow
14577796 1 10 to 100 μL Yellow
14577797 1 20 to 200 μL Yellow
14577798 1 100 to 1000 μL Blue
14577799 1 500 to 5000 μL Green
14577800 1 1000 to 10,000 μL Red
14577801 8 0.5 to 10 μL Gray
14577802 8 5 to 100 μL Yellow
14577803 8 30 to 300 μL Orange
14577804 12 0.5 to 10 μL Gray
14577805 12 5 to 100 μL Yellow
14577806 12 30 to 300 μL Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 14577793 Supplier Sartorius Supplier No. LH729010
Only null left
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
Specifications
Compatible Tips Optifit 10 μL, SafetySpace Filter Tips 10 μL
Adjustable Tip Spacing Yes
Autoclavable Autoclavable
Color Gray
Disposable Reusable
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
Product Line Tacta
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 0.1 to 3 μL
Show More Show Less