missing translation for 'onlineSavingsMsg'
Learn More

Tacrolimus, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111018153
Change view
Click to view available options
Quantity:
150 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018153 150 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018153 Supplier Merck Emd Millipore Supplier No. PHR1809150MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 109581-93-3
Linear Formula C44H69NO12 · H2O
MDL Number MFCD11045918
Synonym FK-506 monohydrate; Tacrolimμs
Quantity 150 mg
Formula Weight 822.03
Grade Certified Reference Material