missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tacrolimus Monohydrate 80.0+%, TCI America™
Supplier: TCI America M225850MG
Specifications
| Tacrolimus Monohydrate | |
| 127°C | |
| C44H71NO13 | |
| 50 mg | |
| FK 506 | |
| CC1CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC(=O)C(C=C(C1)C)CC=C)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC.O | |
| 5282315 | |
| 804.03 | |
| Crystalline Powder |
| 109581-93-3 | |
| White | |
| MFCD11045918 | |
| 2811 | |
| NWJQLQGQZSIBAF-MLAUYUEBSA-N | |
| 822.046 | |
| CHEBI:61057 | |
| ≥80.0% (HPLC) |
Chemical Identifiers
| 109581-93-3 | |
| 822.046 | |
| NWJQLQGQZSIBAF-MLAUYUEBSA-N | |
| 5282315 | |
| CC1CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC(=O)C(C=C(C1)C)CC=C)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC.O |
| C44H71NO13 | |
| MFCD11045918 | |
| FK 506 | |
| CHEBI:61057 |
Safety and Handling
RTECSNumber : KD4201200
TSCA : Yes
Recommended Storage : Freezer