missing translation for 'onlineSavingsMsg'
Learn More

Tacrolimus Monohydrate 80.0+%, TCI America™

Catalog No. M225810MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M225810MG 10 mg
M225850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M225810MG Supplier TCI America Supplier No. M225810MG

Chemical Identifiers

CAS 109581-93-3
Molecular Formula C44H71NO13
Molecular Weight (g/mol) 822.046
MDL Number MFCD11045918
InChI Key NWJQLQGQZSIBAF-MLAUYUEBSA-N
Synonym FK 506
PubChem CID 5282315
ChEBI CHEBI:61057
SMILES CC1CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC(=O)C(C=C(C1)C)CC=C)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC.O

Specifications

Melting Point 127°C
Color White
Quantity 10 mg
UN Number 2811
Formula Weight 804.03
Percent Purity ≥80.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tacrolimus Monohydrate
RTECSNumber KD4201200
TSCA Yes
Recommended Storage Freezer