missing translation for 'onlineSavingsMsg'
Learn More

Metro™ TableWorx Performance Work Table without Backsplash, Mobile-Ready, Standard Series

Catalog No. 01258758
Change view
Click to view available options
Dimensions (L x W):
8 x 36 in.
24 x 24 in.
30 x 24 in.
30 x 30 in.
36 x 24 in.
36 x 30 in.
36 x 36 in.
48 x 24 in.
48 x 30 in.
60 x 24 in.
60 x 30 in.
60 x 36 in.
72 x 24 in.
72 x 30 in.
96 x 24 in.
96 x 30 in.
Frame Type:
3-sided Frame
I-Frame
Undershelf
44 product options available for selection
Product selection table with 44 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Frame Type
01258758 24 x 24 in. Undershelf
01258755 24 x 24 in. I-Frame
01258752 24 x 24 in. 3-sided Frame
01258749 30 x 24 in. Undershelf
01258745 30 x 24 in. I-Frame
01258739 36 x 24 in. Undershelf
01258736 36 x 24 in. I-Frame
01258733 36 x 24 in. 3-sided Frame
01258730 48 x 24 in. Undershelf
01258727 48 x 24 in. I-Frame
01258724 48 x 24 in. 3-sided Frame
01258721 60 x 24 in. Undershelf
01258718 60 x 24 in. I-Frame
01258715 60 x 24 in. 3-sided Frame
01258712 72 x 24 in. Undershelf
01258709 72 x 24 in. I-Frame
01258706 72 x 24 in. 3-sided Frame
01258703 96 x 24 in. Undershelf
01258700 96 x 24 in. I-Frame
01258697 96 x 24 in. 3-sided Frame
01258694 30 x 30 in. Undershelf
01258691 30 x 30 in. I-Frame
01258688 30 x 30 in. 3-sided Frame
01258685 36 x 30 in. Undershelf
01258682 36 x 30 in. I-Frame
01258679 36 x 30 in. 3-sided Frame
01258676 48 x 30 in. Undershelf
01258673 48 x 30 in. I-Frame
01258664 60 x 30 in. I-Frame
01258661 60 x 30 in. 3-sided Frame
01258658 72 x 30 in. Undershelf
01258655 72 x 30 in. I-Frame
01258652 72 x 30 in. 3-sided Frame
01258649 96 x 30 in. Undershelf
01258646 96 x 30 in. I-Frame
01258643 96 x 30 in. 3-sided Frame
01258640 36 x 36 in. Undershelf
01258637 36 x 36 in. I-Frame
01258631 8 x 36 in. Undershelf
01258628 8 x 36 in. I-Frame
01258625 8 x 36 in. 3-sided Frame
01258622 60 x 36 in. Undershelf
01258619 60 x 36 in. I-Frame
01258616 60 x 36 in. 3-sided Frame
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
44 options
Catalog No. 01258758 Supplier Metro™ Supplier No. TWM2424FS304K
Only null left

Metro TableWorx is a truly modular platform of performance work tables designed to allow you to customize your perfect solution. Standard Series TableWorx Tables feature a 304 stainless steel surface and rust-resistant Metroseal Gray epoxy coated legs with built-in Microban antimicrobial protection.

  • All TableWorx tables are available in 3 different frame types: 3-Sided, I-Frame, and Undershelf.
    • A type 304 stainless steel 3-sided frame allows for equipment to be nested underneath the table, and is also ideal for seated use of the work table.
    • A sturdy, type 304 stainless steel I-frame is ideal for applications that require 2-sided access to the work table. It can also be used as a foot rest for seated applications.
    • A solid, type 304 stainless steel undershelf provides additional storage space.
  • Out-of-the-box, TableWorx can be assembled to any one of six working surface heights: 34 in. (864 mm), 35 in. (889 mm), 36 in. (914 mm), 37 in. (940 mm), 38 in. (965 mm), and 39 in. (991 mm).
  • All sizes are available with an optional 2 in. (51 mm) stainless steel backsplash, which is formed into the work surface.
  • 16-gauge, Type 304 stainless steel work surface reinforced with 16-gauge hat channels.
  • The sound deadened work surface features 90-degree turned down edges on all four sides.
  • Stationary work table legs include stainless steel leveling bullet feet with 1.125 in. (29 mm) range of adjustability. Mobile-ready versions are also available, and can accept any Metro 1 in. (25 mm) stem caster. Please note that casters are NOT included with mobile-ready TableWorx models and must be ordered separately.
  • Elevate your efficiency to new levels with the addition of TableWorx productivity riser. Risers can be easily added to any TableWorx work table. A full array of overshelves, wire grids, and accessories can be added to maximize every inch of space, making you more comfortable and productive than ever before. Risers can be mounted at the rear or center of the work table.
  • See Spec Sheet for Additional Information

Specifications

Product Type Work Table
Width (English) 24 in.
Length (English) 24 in.
Dimensions (L x W) 24 x 24 in.
Frame Type Undershelf
Color Silver
Material Stainless Steel