missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAB3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A302207AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
TAB3 functions in the NF-kappaB signal transduction pathway. It and the similar and functionally redundant protein TAB2, form a ternary complex with the protein kinase TAK1 and either TRAF2 or TRAF6 in response to stimulation with the pro-inflammatory cytokines TNF or IL-1. Subsequent TAK1 kinase activity triggers a signaling cascade leading to activation of the NF-kappaB transcription factor. Recent experiments have shown that TAB2 and the related protein TAB3 constitutitvely interact with the autophagy mediator Beclin-1; upon induction of autophagy, these proteins dissociate from Beclin-1 and bind TAK1. Overexpression of TAB2 and TAB3 inhibit autophagy, while their depletion triggers it, suggesting that TAB2 and TAB3 act as a control point for autophagy.Specifications
| TAB3 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q571K4, Q8N5C8 | |
| Tab3 | |
| Between 600 and 650 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| TAB3 | |
| TAB3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 257397, 66724 | |
| 4° C | |
| Liquid |