missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAAR1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA523141

Catalog No. PIPA523141


Only null left

Description

Description

The amino acid sequence used as immunogen is 86% homologous in Mouse and 80% homologous in Canine.

TA1 has been reported in human brain, dorsal root ganglion, olfactory bulb, kidney, liver, lung, pancreas, prostate, skeletal muscle, small intestine, spleen, spinal cord, and stomach. An EST for TA1 has been identified from a human stomach cancer library. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates.
Specifications

Chemical Identifiers

TAAR1
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Monkey
Taar1
MGC126874; MGC138399; RP11-295F4.9; RTAR1; Snorf33; Ta1; Taar1; TAR1; taR-1; trace amine associated receptor 1; trace amine receptor 1; trace amine-associated receptor 1; trace-amine-associated receptor 1; TRAR1
Taar1
Primary
Protein G
1.0 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA and 0.05% sodium azide
Q96RJ0
134864
A portion of amino acids 225-250 of human TAAR1.
Antibody

Specifications

TAAR1
Polyclonal
Unconjugated
Taar1
MGC126874; MGC138399; RP11-295F4.9; RTAR1; Snorf33; Ta1; Taar1; TAR1; taR-1; trace amine associated receptor 1; trace amine receptor 1; trace amine-associated receptor 1; trace-amine-associated receptor 1; TRAR1
Rabbit
Protein G
RUO
134864
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.0 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q96RJ0
Taar1
A portion of amino acids 225-250 of human TAAR1.
100 μg
Primary
Human, Monkey
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions