missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ T98G Cell Lysate

Catalog No. NBP225054
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225054 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225054 Supplier R&D Systems™ Supplier No. NBP225054

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Applications: SDS-Page

The T98G Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.

Specifications

Cell Line T98G cell line lysate (human glioma cell line)
Quantity 0.1 mg
Storage Buffer 1X SDS sample buffer: 60mM Tris-HCl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol
Host Species Human
Content And Storage -80°C in the dark. Avoid freeze-thaw cycles.
Product Line Novus

For Research Use Only