Learn More
G-Biosciences T7 RNA Polymerase
A DNA-dependent RNA polymerase that is highly specific for the T7 phage promoters.
$170.54 - $709.23
Description
T7 RNA Polymerase catalyzes in vitro RNA synthesis from a cloned DNA sequence under the T7 promoters. It has a wide range of applications including Preparation of RNA vaccines, Radiolabeled RNA probes, Guide RNA for gene targeting. mRNA for in vitro translation and micro injection, RNA amplification.
Specifications
| T7 RNA Polymerase | |
| TheraPure™ GMP | |
| T7 |