Learn More
G-Biosciences T7 RNA Polymerase
Catalog No. S000753
Encompass
Change view
Click to view available options
Quantity:
25000 U
5000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000753 25000 U
S000747 5000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000753 Supplier G-Biosciences Supplier No. 7862011
Only null left
A DNA-dependent RNA polymerase that is highly specific for the T7 phage promoters.
T7 RNA Polymerase catalyzes in vitro RNA synthesis from a cloned DNA sequence under the T7 promoters. It has a wide range of applications including Preparation of RNA vaccines, Radiolabeled RNA probes, Guide RNA for gene targeting. mRNA for in vitro translation and micro injection, RNA amplification.
Specifications
Polymerase T7 RNA Polymerase
Product Line TheraPure™ GMP
Promoter T7
Quantity 25000 U