missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ T.T.C. Solution 5%
Description
Oxoid TTC Solution is used for the enumeration and identification of coliform bacteria.
Specifications
Specifications
| Format | Liquid |
| Product Type | Microbiology Media |
| Quantity | 1 x 100 mL |
| Yield | As required |