missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ T-Shaped Connecting Tube
For use with flexible tubes and/or hoses.
$40.20 - $44.63
Specifications
| Material | Borosilicate Glass |
|---|---|
| For Use With | Flexible Tubing |
| For Use With (Application) | Organic Synthesis |
| Packaging Quantity | 1 Ea. |
| Color | Clear |
Specifications
| Borosilicate Glass | |
| Organic Synthesis | |
| Clear |
| Flexible Tubing | |
| 1 Ea. | |
| T-Type Connecting Tube |