missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ T-Shaped Connecting Tube
For use with flexible tubes and/or hoses.
Supplier: Synthware™ T120800
Specifications
| Borosilicate Glass | |
| 8 mm | |
| Organic Synthesis | |
| 8 mm | |
| T-Type Connecting Tube |
| 0.32 in. | |
| Flexible Tubing | |
| 1 Ea. | |
| Clear |