missing translation for 'onlineSavingsMsg'
Learn More

T Cell Receptor alpha Chain-V alpha 24-J alpha Q Antibody [DyLight 405], R&D Systems™

Catalog No. A40008171
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40008171 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40008171 Supplier Thermo Fisher Scientific Supplier No. A40008171
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

T Cell Receptor alpha Chain-V alpha 24-J alpha Q Polyclonal specifically detects T Cell Receptor alpha Chain-V alpha 24-J alpha Q in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen T Cell Receptor alpha Chain-V alpha 24-J alpha Q
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Host Species Sheep
Immunogen E. coli-derived recombinant human T Cell Receptor alpha Chain-V alpha 24-J alpha Q, Lys22-Pro135, Accession # ABC72391
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less