missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTOX™ Deep Red Nucleic Acid Stain, for fixed/dead cells

Catalog No. p-7180612
Change view
Click to view available options
Quantity:
50 μL
5 x 50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11381 5 x 50 μL
S11380 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S11381 Supplier Invitrogen™ Supplier No. S11381
Only null left

SYTOX Deep Red Nucleic Acid Stain is a high-affinity nucleic acid stain that easily penetrates cells with compromised plasma membranes and yet does not cross the membranes of live cells.

With minimal cytoplasmic staining, SYTOX Deep Red stain is particularly useful in immunocytochemistry (ICC), immunohistochemistry (IHC), and immunofluorescence (IF) experiments for staining of nuclei of fixed cells. After a brief incubation with SYTOX Deep Red stain, the nucleic acids of dead or fixed cells fluoresce a bright, deep red/far red and can be detected with a CY5/Deep Red standard filter set or laser configuration.

Features of SYTOX Deep Red Nucleic Acid Stain include:
• Impermeant to live cells; permeant to dead or fixed cells
• Excitation/emission peak: 660/682 nm
• Use with Cy5 traditional filter set or 647 laser line
• Works with mammalian cells in mono layer or 3D cell culture, animal tissue, and bacteria
• Detectable with fluorescence micro¬scopes, fluorimeters, fluorescence microplate readers, and flow cytometers

SYTOX Deep Red Nucleic Acid Stain is multiplexable with blue, green, orange, red, and near IR fluorophores, when compatible fluorescent filter/laser configurations are used. SYTOX Deep Red stain shows increased fluorescence with increasing concentrations of dsDNA, but does not show much affinity for RNA or ssDNA. It has a bright initial signal and excellent photo stability in a typical imaging experiment. These properties make SYTOX Deep Red stain a simple and quantitative single-step dead/fixed cell nucleus-labeling dye for use with fluorescence micro¬scopes, fluorimeters, fluorescence microplate readers, and flow cytometers.

SYTOX Deep Red Nucleic Acid Stain had been used successfully in mono-layer cells, thin-sliced tissue, thick-sliced tissue, 3D cell cultures/spheroids, and bacteria. It is supplied at a 1 mM concentration in DMSO (∼2000X concentration). A concentration of 0.5 μM (1X) is recommended for most applications and cell types. In fluorescent imaging of various mammalian cells, a concentration range of 2.5 μM to 0.25 μM has been used. For viability testing by flow cytometry, a concentration of 1 μM to 10 nM has been used for Jurkat cells and a range of 4 μM to 0.5 μM has been used with E. coli.

Specifications

Color Deep Red
Content And Storage Store in freezer at -5°C to -30°C and protect from light.
Excitation Wavelength Range 660 nm
Dye Type Cell-Permeant
For Use With (Application) Viability Assay
For Use With (Equipment) Fluorescence Microscope
Quantity 5 x 50 μL
Detection Method Fluorescence
Emission 682 nm
Product Line SYTOX
Shipping Condition Approved for shipment on Wet or Dry Ice
Product Type Nucleic Acid Stain
SubCellular Localization Nucleic Acids
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.