Learn More
Invitrogen™ SYTOX™ Deep Red Nucleic Acid Stain, for fixed/dead cells
Description
With minimal cytoplasmic staining, SYTOX Deep Red stain is particularly useful in immunocytochemistry (ICC), immunohistochemistry (IHC), and immunofluorescence (IF) experiments for staining of nuclei of fixed cells. After a brief incubation with SYTOX Deep Red stain, the nucleic acids of dead or fixed cells fluoresce a bright, deep red/far red and can be detected with a CY5/Deep Red standard filter set or laser configuration.
Features of SYTOX Deep Red Nucleic Acid Stain include:
• Impermeant to live cells; permeant to dead or fixed cells
• Excitation/emission peak: 660/682 nm
• Use with Cy5 traditional filter set or 647 laser line
• Works with mammalian cells in mono layer or 3D cell culture, animal tissue, and bacteria
• Detectable with fluorescence micro¬scopes, fluorimeters, fluorescence microplate readers, and flow cytometers
SYTOX Deep Red Nucleic Acid Stain is multiplexable with blue, green, orange, red, and near IR fluorophores, when compatible fluorescent filter/laser configurations are used. SYTOX Deep Red stain shows increased fluorescence with increasing concentrations of dsDNA, but does not show much affinity for RNA or ssDNA. It has a bright initial signal and excellent photo stability in a typical imaging experiment. These properties make SYTOX Deep Red stain a simple and quantitative single-step dead/fixed cell nucleus-labeling dye for use with fluorescence micro¬scopes, fluorimeters, fluorescence microplate readers, and flow cytometers.
SYTOX Deep Red Nucleic Acid Stain had been used successfully in mono-layer cells, thin-sliced tissue, thick-sliced tissue, 3D cell cultures/spheroids, and bacteria. It is supplied at a 1 mM concentration in DMSO (∼2000X concentration). A concentration of 0.5 μM (1X) is recommended for most applications and cell types. In fluorescent imaging of various mammalian cells, a concentration range of 2.5 μM to 0.25 μM has been used. For viability testing by flow cytometry, a concentration of 1 μM to 10 nM has been used for Jurkat cells and a range of 4 μM to 0.5 μM has been used with E. coli.
Specifications
Specifications
| Color | Deep Red |
| Content And Storage | Store in freezer at -5°C to -30°C and protect from light. |
| Excitation Wavelength Range | 660 nm |
| Dye Type | Cell-Permeant |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 50 μL |
| Volume (Metric) | 50 μL |
| Detection Method | Fluorescence |
| Emission | 682 nm |
| Show More |
Frequently Asked Questions (FAQs)
Yes. For live cell staining, you may use any of the blue, green, or orange SYTO dyes or any of the blue fluorescent Hoechst stains.
We do not recommend using SYTOX Deep Red Nucleic Acid Stain for cell cycle analysis. Instead, we recommend using Vybrant DyeCycle Ruby Stain (Cat Nos. V10273, V10309).
Although they are both far red, cell impermeant nucleic acid stains, there is approximately a 20 nm difference in the excitation and emission maxima: SYTOX Deep Red Nucleic Acid Stain ex/em = 660/682 nm and SYTOX Red Dead Cell Stain ex/em = 640/658. In addition, the main difference is that SYTOX Deep Red Nucleic Acid Stain has less propensity to stain RNA compared to SYTOX Red Dead Cell Stain.
For Research Use Only. Not for use in diagnostic procedures.