missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ SYTOX™ AADvanced™ Dead Cell Stain Kit

Catalog No. p-7076272
Encompass
Change view
Click to view available options
Quantity:
100 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121541 100 Reactions
501137611 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501121541 Supplier Molecular Probes™ Supplier No. S10349
Only null left

SYTOX™ AADvanced™ Dead Cell Stain is a new high-affinity nucleic acid stain for the detection of dead cells and analysis of cell cycle using the common 488 nm blue laser in flow cytometry.

  • A new high-affinity nucleic acid stain for the detection of dead cells and analysis of cell cycle using the common 488 nm blue laser in flow cytometry
  • The dye is spectrally similar to 7-AAD but with rapid kinetics of uptake and relatively low CVs
  • SYTOX™ AADvanced™ Dead Cell Stain penetrates cells more efficiently than 7-AAD providing better separation of live and dead cells
  • It can also be used with fixed cells for DNA content analysis when paired with RNAse treatment
  • Kits contain a vial(s) of dried dye and anhydrous DMSO providing stable shelf life

Specifications

Cell Permeability Impermeant
Color Red
Content And Storage Contains 1 vial of SYTOX™ AADvanced™ dead cell stain and 200 μL dimethylsulfoxide (DMSO).

Store at ≤-20°C and protect from light.
Description SYTOX™ AADvanced™ Dead Cell Stain Kit
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dead Cell Stain Kit
Dye Type SYTOX™ AADvanced™
Emission 647
Excitation Wavelength Range 546 nm
Form Solid
Format Tube(s)
Product Line Molecular Probes, SYTOX
Quantity 100 Reactions
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Sub Cellular Localization Nucleus, Nucleic Acids
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.