missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTO™ RNASelect™ Red

Catalog No. S32711
Change view
Click to view available options
Quantity:
300 μL
3 x 300 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S32711 3 x 300 μL
S32710 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S32711 Supplier Invitrogen™ Supplier No. S32711

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SYTO™ RNASelect™ Red is a cell-permeant, RNA-selective fluorescent dye for live or fixed eukaryotic cells. It allows researchers to visualize RNA-rich nucleoli and cytoplasmic RNA in a Texas Red/RFP compatible channel.

SYTO™ RNASelect™ Red is a cell-permeant, RNA-selective fluorescent dye for live or fixed eukaryotic cells. It allows researchers to visualize RNA-rich nucleoli and cytoplasmic RNA in a Texas Red/RFP compatible channel. Compared to SYTO™ RNASelect™ Green, it is compatible with 4% paraformaldehyde fixation rather than methanol fixation, and leaves GFP/FITC channels open for antibodies, fluorescent proteins, and other fluorescent probes in multiplex and high-content imaging.

This molecular probe is prepared as a ready-to-use 100x aqueous solution, appropriate for direct addition to either complete media or other buffers of your choice to then be used in staining cells.

Compatibility with 4% paraformaldehyde fixation and permeabilization expands the applicability of this RNA dye for immunofluorescence workflows, enabling its use in high content screening for nucleolar reorganization, cell stress assays, and deep learning computational models.

The staining patterns demonstrate high RNA specificity, with expected staining demonstrating bright nucleoli and diffuse ribosomal staining throughout the cytoplasm and is easily multiplexable with classic nuclear stains such as Hoechst 33342 or DAPI. This specificity is demonstrated by the sharp decrease in staining intensity upon treatment with RNase, all red signal is quickly depleted while the nuclear DNA dye staining remains intact.

Features

  • Suitable for both live and fixed cell workflows, connecting live cell observation with endpoint imaging.
  • Compatible with multiplex imaging outside of the Texas Red/RFP channels with various organelle markers such as Hoechst/DAPI, phalloidin, ER, lectins, or mitochondrial probes.
  • Applicable to high-content imaging and RNA based nucleolar morphology studies, including workflows where nucleolar signal is used for segmentation or comparative phenotyping.

For researchers familiar with SYTO™ 14, SYTO™ RNASelect™ Red offers a red channel option for RNA associated morphology in Cell Painting related or other multiplex imaging workflows.

This dye complements SYTO™ RNASelect™ Green in channel planning and fixation choice: SYTO™ RNASelect™ Green remains the better fit when the green channel is desired and the sample type requires methanol fixation, whereas SYTO™ RNASelect™ Red is appropriate when a red RNA channel or 4% PFA workflow is preferred.

Available as a single vial as S32710 and in a 3-pack format as S32711.

Specifications

Color Red
Concentration 1 mM in 25% DMSO/DI H2O
Content And Storage •3 Vials
•Store at 2°C to 30°C
•Protect from light
•Do not freeze
Excitation Wavelength Range 595 nm
Dye Type Fluorogenic Organic Dye
Format Vial
For Use With (Application) Live Cell Imaging, Fixed Cell Imaging, High Content Screening, Fluorescence Microscopy
For Use With (Equipment) Fluorescence Microscope, Spectral Imaging Instrument, Flow Cytometer, High Content Imager
No. of Tests 900 Tests
Reactivity RNA, Nucleolar Structure
Quantity 3 x 300 μL
Validated Application Live Cell Imaging, Fixed Cell Imaging, High Content Imaging
Volume (Metric) 900 μL
Detection Method Fluorescence with Texas Red Filter Set
Emission 626 nm
Form Aqueous Liquid with 25% DMSO
Product Line SYTO RNASelect
Reagent Type Fluorogenic RNA Label
Target RNA
Technique High Content Imaging, Fluorescence Microscopy
Label Type Fluorescent Dye
Product Type RNA-specific Label
Sub Cellular Localization Nucleolus
Cell Permeability Live and Fixed Cell Permeable
Show More Show Less

For Research Use Only.