missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTO™ Green Fluorescent Nucleic Acid Stains

Catalog No. s32703
Encompass
Change view
Click to view available options
Dye Type:
SYTO 11 Green
SYTO 12 Green
SYTO 13 Green
SYTO 14 Green
SYTO 16 Green
SYTO 21 Green
SYTO 24 Green
SYTO 9 Green
SYTO BC Green
SYTO Green Dyes 11-14, 16, 21, 24, and 25
SYTO RNASelect Green
Quantity:
1 Kit (50 μL each)
100 μL
250 μL
315 μL
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dye Type Quantity
S32703 SYTO RNASelect Green 100 μL
S34854 SYTO 9 Green 100 μL
S7573 SYTO 11 Green 250 μL
S7574 SYTO 12 Green 315 μL
S7575 SYTO 13 Green 250 μL
S7576 SYTO 14 Green 250 μL
S7578 SYTO 16 Green 250 μL
S7556 SYTO 21 Green 250 μL
S7559 SYTO 24 Green 250 μL
501121577 SYTO BC Green 100 μL
S7572 SYTO Green Dyes 11-14, 16, 21, 24, and 25 1 Kit (50 μL each)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. S32703 Supplier Invitrogen™ Supplier No. S32703

SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria.

The SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria. Because the dye characteristics and staining pattern of the SYTO dyes may vary, we offer 10 different SYTO Green Fluorescent Nucleic Acid Stains available individually along with the SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 (containing SYTO 11–14, 16, 21, 24, and 25) to enable researchers to find the most appropriate green-fluorescent SYTO stain for their system . We also provide the SYTOX Green Nucleic Acid Stain (Cat. No. S7020 and R37109), which is a cell-impermeant green-fluorescent stain used for staining dead cells or performing nuclear counterstaining in fixed cells.

SYTO Green Fluorescent Nucleic Acid Stains are cell-permeant nucleic acid stains that show a large fluorescence enhancement upon binding nucleic acids. The SYTO dyes can be used to stain RNA and DNA in both live and dead eukaryotic cells, as well as in Gram-positive and Gram-negative bacteria.

The SYTO stains share several important characteristics:

  • Permeability to virtually all cell membranes, including mammalian cells and bacteria
  • High molar absorptivity, with extinction coefficients >50,000 cm-1 M-1 at visible absorption maxima
  • Extremely low intrinsic fluorescence, with quantum yields typically <0.01 when not bound to nucleic acids
  • Quantum yields that are typically >0.4 when bound to nucleic acids.

SYTO Green-Fluorescent Nucleic Acid Stains excitation/emission wavelengths

  • SYTO 9—RNA 486/501 nm; DNA 485/498 nm
  • SYTO 11—RNA 510/530 nm; DNA 508/527 nm
  • SYTO 12—RNA 500/519 nm; DNA 499/522 nm
  • SYTO 13—RNA 491/514 nm; DNA 488/509 nm
  • SYTO 14—RNA 521/547 nm; DNA 517/549 nm
  • SYTO 16—RNA 494/525 nm; DNA 488/518 nm
  • SYTO 21—DNA 494/517 nm
  • SYTO 24—DNA 490/515 nm
  • SYTO BC—RNA 487/504 nm; DNA 485/500 nm
  • SYTO RNASelect—RNA 490/530 nm

SYTO dyes differ from each other in one or more characteristics, including cell permeability, fluorescence enhancement upon binding nucleic acids, excitation and emission spectra, DNA/RNA selectivity, and binding affinity. The SYTO dyes are compatible with a variety of fluorescence-based instruments that use laser excitation or a conventional broadband illumination source (e.g., mercury- and xenon-arc lamps). Filter sets that are suitable for imaging cells labeled with fluorescein (FITC), Alexa Fluor 488, or GFP will work well for imaging cells stained with SYTO Green Nucleic Acid Stains.

SYTO nucleic acid stains have been used in diverse applications from staining DNA spotted on microarrays to staining live and fixed cells. The SYTO dyes do not act exclusively as nuclear stains in live cells and should not be equated with DNA-selective compounds such as DAPI (Cat. Nos. D1306, D21490) or Hoechst 33342 (Cat. Nos. H1399, H3570), which stain nuclei in live animal cells. Eukaryotic cells incubated with SYTO dyes generally show cytoplasmic or mitochondrial staining, as well as nuclear staining. The SYTO Green Fluorescent Nucleic Acid Stains have proven valuable in a broad range of research applications as well as used in flow cytometry and fluorescence microscopy.

Specific staining applications for some of the SYTO Green Fluorescent Nucleic Acid Stains

The SYTO 9 stain has been shown to stain live and dead Gram-positive and Gram-negative bacteria. It is a component of the LIVE/DEAD BacLight Bacterial Viability Kits (Cat. Nos. L7007, L7012, L13152).

The SYTO 11 stain (Cat. No. S7573) has been used in conjunction with time-lapse microscopy to examine the cleavage orientation of dividing cells in the developing cerebral cortex.

The SYTO 13 stain (Cat. No. S7575) has been used in combination with propidium iodide (Cat. Nos. P1304, P3566) to monitor glutamate-induced necrosis in cerebellar granule cells.

The SYTO 14 stain (Cat. No. S7576) binds to cytoplasmic RNA, allowing its use in tracking RNA granule transport in living neurons.

Several reports describe the use of SYTO dyes for detecting apoptosis. A series of SYTO nucleic acid stains was screened for the ability to discriminate between apoptotic and non-apoptotic mouse thymocytes, and the SYTO 16 stain (Cat. no. S7578) was found to be optimal for this application. The SYTO 16 stain has also been used with propidium iodide to differentiate live and dead COS-7 cells with a laser-based scanning cytometer.

The SYTO BC Green Fluorescent Nucleic Acid Stain is a mixture of some of our best SYTO dyes for bacterial staining and is a component of the Bacteria Counting Kit (Cat. No. B7277) for bacterial counting in flow cytometry.

The SYTO RNASelect Green Fluorescent Cell Stain is a cell-permeant nucleic acid stain that selectively stains RNA. Although virtually nonfluorescent in the absence of nucleic acids, the SYTO RNASelect stain exhibits bright green fluorescence when bound to RNA (absorption/emission maxima ∼490/530 nm), but only a weak fluorescent signal when bound to DNA. It has been used for staining RNA cargo in exosomes in both live cell samples and isolated exosomes.

SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit

The SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 contains our collection of cell-permeant, green-fluorescent SYTO nucleic acid stains. Because the dyes may demonstrate different staining behaviors with various tissues and cells, it may be necessary to test the dyes to find the optimal dye for a specific application. The kit contains 50 μL each of SYTO 11–14, 16, 21, 24, and 25 dyes.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

Order Info

Shipping Condition: Room Temperature

Specifications

Color Green
Concentration 5 mM solution in DMSO
Content And Storage Store in freezer at -5°C to -30°C and protect from light.
Excitation Wavelength Range RNA: 490 nm
Dye Type SYTO RNASelect Green
For Use With (Application) Fluorescent Labeling, Live Cell Imaging, Microarray
For Use With (Equipment) Fluorescence Microscope, High Content Imager, Flow Cytometer
Storage Buffer DMSO
Quantity 100 μL
Volume (Metric) 100 μL
Detection Method Fluorescence
Emission RNA: 530 nm
Product Line SYTO
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Nucleic Acid Stain
Sub Cellular Localization Nucleic Acids
Show More Show Less
How do SYTO dyes bind to DNA?

The binding mode of SYTO nucleic acid stains is unknown. However, the behavior of these and related nucleic acid dyes suggests the following binding properties:

1.They appear to contact the solvent (suggested by sensitivity to salt, divalent cations, and in particular, SDS) and thus are likely to have contacts in the grooves.
2.All SYTO dyes appear to show some base selectivity and are thus likely to have minor groove contacts.
3.They can be removed from nucleic acid via ethanol precipitation; this characteristic is not shared by ethidium bromide and other intercalators. Likewise, the dyes are not removed from nucleic acid via butanol or chloroform extraction. These extraction methods do remove ethidium bromide from nucleic acid. 4. SYTO binding is not affected by nonionic detergents.
5. SYTO dyes are not quenched by BrdU, so they do not bind nucleic acids in precisely the same way as Hoechst 33342 and DAPI ((4′,6-diamidino-2-phenylindole).

SYBR Green I has shown little mutagenicity on frameshift indicator strains, indicating that it isn't likely to strongly intercalate.

Are your exosome kits for the isolation of exosomes from serum species-specific?

The reagent for exosome isolation from serum can be used with serum from any species in addition to human. When working with small volumes of non-human samples (e.g., mouse) or when it is desired to maximize the recovery of exosomes, we recommend following the standard protocol except for spinning the samples (after incubation with the reagent) to precipitate the exosomes at 4 degrees C.

Can I use PBS with SYTO RNASelect Red, SYTO RNASelect Green, and SYTO 14 reagents?

PBS can be acceptable in some workflows, but we do not recommend assuming it is always the best-performing buffer for all three probes. Some applications may perform better in phosphate-free buffers, so we suggest optimizing as needed. When a starting point is needed, we recommend Live Cell Imaging Solution (LCIS; Cat. No. A59688DJ).

Can I use SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents as absolute measurements of total RNA content?

We recommend using these stains as relative imaging reagents within a controlled workflow rather than as absolute measurements of total RNA content. SYTO RNASelect Green reagent has been used in quantitative RNA content assay workflows, but any quantitative application should be carefully calibrated and ideally confirmed with an orthogonal RNA quantitation method.

Are SYTO RNASelect Red, SYTO RNASelect Green, and SYTO 14 reagents transcript-specific?

No. We recommend thinking of SYTO RNASelect Red, SYTO RNASelect Green, and SYTO 14 reagent as RNA-associated stains rather than sequence-targeted probes.

Can I use SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents for automated segmentation or machine-learning analysis?

Yes. We suggest these dyes can be used for automated segmentation and machine-learning analysis, depending on the workflow. SYTO 14 reagent has strong Cell Painting precedent, and SYTO RNASelect Red reagent has shown utility in high-content imaging workflows that rely on segmentation-oriented nucleolar analysis.

Why is my nucleolar signal weak with SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents?

We recommend optimizing the staining protocol by increasing the dye concentration stepwise, confirming that the stock is fully homogeneous, extending the incubation time within the documented range, and verifying that the fixation method matches the dye. For SYTO RNASelect Green reagent, we do not recommend PFA fixation because it can flatten the expected staining pattern.

Why is my background high with SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents?

We recommend first reducing the dye concentration and shortening the incubation time. We also suggest adding a wash step, comparing your culture medium with a low-background imaging buffer, and confirming that the stock solution was fully dissolved before dilution.

Why did the punctate mitochondrial signal disappear after methanol fixation with SYTO RNASelect Green reagent?

We suggest interpreting this as a fixation-related change in staining pattern. After methanol fixation, RNA-associated staining can be preserved while the punctate mitochondrial-like signal diminishes. Because these dyes are cationic, healthy mitochondria can contribute to that punctate signal in live cells, and that feature may be reduced after fixation.

Why is my SYTO RNASelect Green reagent signal more diffuse after fixation?

If PFA was used, redistribution of the dye is the most likely cause. We recommend methanol fixation rather than PFA for SYTO RNASelect Green reagent.

Why is my SYTO RNASelect Green reagent working solution leaving debris on the coverslip?

This is often caused by precipitation in the working solution. We recommend preparing SYTO RNASelect Green reagent fresh with pre-warmed LCIS and using it immediately, because the diluted dye can begin to precipitate if it sits for more than about 2–5 minutes.

Why should I use plastic tubes for SYTO dye dilutions?

We recommend using plastic tubes for dilutions because SYTO dyes can adhere to glass, which can lower the effective concentration and increase variability.

Do I need to warm frozen SYTO 14 or SYTO RNASelect Green reagents before opening the vial?

Yes. Both SYTO RNASelect Green and SYTO 14 reagents are stored at -25°C to -5°C. Because these products contain 100% DMSO and are frozen, we recommend letting the vial warm to room temperature before opening it and then briefly centrifuging it before use.

Can I use SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents with fluorescent protein reporters?

Yes, but we recommend choosing the RNA dye around the existing reporter. For example, if GFP is already present, we usually suggest SYTO RNASelect Red reagent to keep the green channel available. During assay setup, we also recommend checking the RFP channel for any bleed-through from the SYTO dye.

Can I use more than one of SYTO RNASelect Red, SYTO RNASelect Green, and SYTO 14 reagents on the same sample?

We do not recommend this for routine imaging. Because SYTO RNASelect Red, SYTO RNASelect Green, and SYTO 14 reagents all bind RNA-associated structures, combining them can create interpretation and competition issues without adding useful biological context.

Can I combine SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents with phalloidin, ER markers, or mitochondrial markers?

Yes. We suggest confirming that the fluorophores are spectrally separated in your imaging setup before finalizing the assay.

Can I combine SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents with DAPI or Hoechst?

Yes. We generally expect good spectral separation, but we recommend matching DAPI or Hoechst to your live-cell or fixed-cell workflow.

Can I combine SYTO RNASelect Green reagent with red-orange fluorophores?

This combination may require optimization. SYTO RNASelect Green reagent can overlap with standard TRITC settings, so we generally suggest avoiding red-orange fluorophores unless bleed-through is acceptable in your assay.

Can I use paraformaldehyde (PFA) with SYTO RNASelect Green reagent?

No. We do not recommend PFA fixation with SYTO RNASelect Green reagent because it can alter the staining pattern and increase cytoplasmic and nuclear background.

Can I stain cells with SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents after fixation?

Yes. We have found that SYTO RNASelect Green, SYTO 14, and SYTO RNASelect Red reagents can all be used after fixation. We do recommend keeping in mind that some staining features can change after fixation. For example, mitochondrial puncta seen with SYTO RNASelect Green reagent may disappear after fixation. SYTO RNASelect Red and SYTO 14 reagents are also compatible with permeabilization before labeling.

Can I stain cells with SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents before fixation?

Yes. We have found that SYTO RNASelect Green, SYTO 14, and SYTO RNASelect Red reagent can all be used to label live cells before fixation. Of the three dyes, only SYTO RNASelect Red and SYTO 14 reagents are compatible with a permeabilization step.

Can I combine SYTO RNASelect Red, SYTO RNASelect Green, or SYTO 14 reagents with antibody staining?

Yes, but the fixation chemistry matters. We recommend SYTO RNASelect Red and SYTO 14 reagents for workflows that use PFA fixation followed by immunofluorescence. SYTO RNASelect Green reagent uses methanol fixation, which can work well for some antibodies but may disrupt epitopes for others.

Can I directly compare signal intensity between SYTO 14, SYTO RNASelect Green, and SYTO RNASelect Red reagents?

We do not recommend treating direct intensity comparisons across SYTO 14, SYTO RNASelect Green, and SYTO RNASelect Red reagents as a simple measure of RNA abundance. These dyes differ in spectra, formulation, brightness, and workflow, so any cross-dye comparison should be considered assay-specific and calibration-dependent.

Which SYTO dye should I choose if my assay already uses GFP or FITC-labeled antibodies?

We usually recommend SYTO RNASelect Red reagent because it keeps the green channel available for GFP or FITC-labeled probes and antibodies.

Which SYTO dye should I choose for a Cell Painting workflow?

We recommend SYTO 14 reagent if you need the closest fit to a Cell Painting workflow because it has the strongest direct literature association with Cell Painting. We suggest using SYTO RNASelect Red reagent as an RNA-focused or channel-specific alternative rather than as a direct replacement for a Cell Painting protocol. We also offer seminar and application note materials describing modified Cell Painting workflows that open channels for additional probes or antibodies.

Which SYTO dye should I choose if I need a green RNA channel?

We recommend SYTO RNASelect Green reagent when methanol fixation is acceptable and a defined RNA-selective workflow is desired. SYTO 14 reagent can also be imaged in FITC/GFP filter sets.

Which SYTO dye should I choose if I need 4% paraformaldehyde (PFA) fixation?

We recommend starting with SYTO RNASelect Red reagent or SYTO 14 if your workflow requires 4% PFA fixation. We recommend cold methanol fixation for SYTO RNASelect Green reagent over PFA fixation.


For Research Use Only. Not for use in diagnostic procedures.