missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTO™ Green Fluorescent Nucleic Acid Stains

Catalog No. S7574
Encompass
Change view
Click to view available options
Dye Type:
SYTO 11 Green
SYTO 12 Green
SYTO 13 Green
SYTO 14 Green
SYTO 16 Green
SYTO 21 Green
SYTO 24 Green
SYTO 9 Green
SYTO BC Green
SYTO Green Dyes 11-14, 16, 21, 24, and 25
SYTO RNASelect Green
Quantity:
1 Kit (50 μL each)
100 μL
250 μL
315 μL
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dye Type Quantity
S7574 SYTO 12 Green 315 μL
S34854 SYTO 9 Green 100 μL
S7573 SYTO 11 Green 250 μL
S7575 SYTO 13 Green 250 μL
S7576 SYTO 14 Green 250 μL
S7578 SYTO 16 Green 250 μL
S7556 SYTO 21 Green 250 μL
S7559 SYTO 24 Green 250 μL
501121577 SYTO BC Green 100 μL
S32703 SYTO RNASelect Green 100 μL
S7572 SYTO Green Dyes 11-14, 16, 21, 24, and 25 1 Kit (50 μL each)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. S7574 Supplier Invitrogen™ Supplier No. S7574

missing translation for 'validMainframeMsg'

SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria.

The SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria. Because the dye characteristics and staining pattern of the SYTO dyes may vary, we offer 10 different SYTO Green Fluorescent Nucleic Acid Stains available individually along with the SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 (containing SYTO 11–14, 16, 21, 24, and 25) to enable researchers to find the most appropriate green-fluorescent SYTO stain for their system . We also provide the SYTOX Green Nucleic Acid Stain (Cat. No. S7020 and R37109), which is a cell-impermeant green-fluorescent stain used for staining dead cells or performing nuclear counterstaining in fixed cells.

SYTO Green Fluorescent Nucleic Acid Stains are cell-permeant nucleic acid stains that show a large fluorescence enhancement upon binding nucleic acids. The SYTO dyes can be used to stain RNA and DNA in both live and dead eukaryotic cells, as well as in Gram-positive and Gram-negative bacteria.

The SYTO stains share several important characteristics:

  • Permeability to virtually all cell membranes, including mammalian cells and bacteria
  • High molar absorptivity, with extinction coefficients >50,000 cm-1 M-1 at visible absorption maxima
  • Extremely low intrinsic fluorescence, with quantum yields typically <0.01 when not bound to nucleic acids
  • Quantum yields that are typically >0.4 when bound to nucleic acids.

SYTO Green-Fluorescent Nucleic Acid Stains excitation/emission wavelengths

  • SYTO 9—RNA 486/501 nm; DNA 485/498 nm
  • SYTO 11—RNA 510/530 nm; DNA 508/527 nm
  • SYTO 12—RNA 500/519 nm; DNA 499/522 nm
  • SYTO 13—RNA 491/514 nm; DNA 488/509 nm
  • SYTO 14—RNA 521/547 nm; DNA 517/549 nm
  • SYTO 16—RNA 494/525 nm; DNA 488/518 nm
  • SYTO 21—DNA 494/517 nm
  • SYTO 24—DNA 490/515 nm
  • SYTO BC—RNA 487/504 nm; DNA 485/500 nm
  • SYTO RNASelect—RNA 490/530 nm

SYTO dyes differ from each other in one or more characteristics, including cell permeability, fluorescence enhancement upon binding nucleic acids, excitation and emission spectra, DNA/RNA selectivity, and binding affinity. The SYTO dyes are compatible with a variety of fluorescence-based instruments that use laser excitation or a conventional broadband illumination source (e.g., mercury- and xenon-arc lamps). Filter sets that are suitable for imaging cells labeled with fluorescein (FITC), Alexa Fluor 488, or GFP will work well for imaging cells stained with SYTO Green Nucleic Acid Stains.

SYTO nucleic acid stains have been used in diverse applications from staining DNA spotted on microarrays to staining live and fixed cells. The SYTO dyes do not act exclusively as nuclear stains in live cells and should not be equated with DNA-selective compounds such as DAPI (Cat. Nos. D1306, D21490) or Hoechst 33342 (Cat. Nos. H1399, H3570), which stain nuclei in live animal cells. Eukaryotic cells incubated with SYTO dyes generally show cytoplasmic or mitochondrial staining, as well as nuclear staining. The SYTO Green Fluorescent Nucleic Acid Stains have proven valuable in a broad range of research applications as well as used in flow cytometry and fluorescence microscopy.

Specific staining applications for some of the SYTO Green Fluorescent Nucleic Acid Stains

The SYTO 9 stain has been shown to stain live and dead Gram-positive and Gram-negative bacteria. It is a component of the LIVE/DEAD BacLight Bacterial Viability Kits (Cat. Nos. L7007, L7012, L13152).

The SYTO 11 stain (Cat. No. S7573) has been used in conjunction with time-lapse microscopy to examine the cleavage orientation of dividing cells in the developing cerebral cortex.

The SYTO 13 stain (Cat. No. S7575) has been used in combination with propidium iodide (Cat. Nos. P1304, P3566) to monitor glutamate-induced necrosis in cerebellar granule cells.

The SYTO 14 stain (Cat. No. S7576) binds to cytoplasmic RNA, allowing its use in tracking RNA granule transport in living neurons.

Several reports describe the use of SYTO dyes for detecting apoptosis. A series of SYTO nucleic acid stains was screened for the ability to discriminate between apoptotic and non-apoptotic mouse thymocytes, and the SYTO 16 stain (Cat. no. S7578) was found to be optimal for this application. The SYTO 16 stain has also been used with propidium iodide to differentiate live and dead COS-7 cells with a laser-based scanning cytometer.

The SYTO BC Green Fluorescent Nucleic Acid Stain is a mixture of some of our best SYTO dyes for bacterial staining and is a component of the Bacteria Counting Kit (Cat. No. B7277) for bacterial counting in flow cytometry.

The SYTO RNASelect Green Fluorescent Cell Stain is a cell-permeant nucleic acid stain that selectively stains RNA. Although virtually nonfluorescent in the absence of nucleic acids, the SYTO RNASelect stain exhibits bright green fluorescence when bound to RNA (absorption/emission maxima ∼490/530 nm), but only a weak fluorescent signal when bound to DNA. It has been used for staining RNA cargo in exosomes in both live cell samples and isolated exosomes.

SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit

The SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 contains our collection of cell-permeant, green-fluorescent SYTO nucleic acid stains. Because the dyes may demonstrate different staining behaviors with various tissues and cells, it may be necessary to test the dyes to find the optimal dye for a specific application. The kit contains 50 μL each of SYTO 11–14, 16, 21, 24, and 25 dyes.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

Order Info

Shipping Condition: Room Temperature

Specifications

Color Green
Concentration 4 mM solution in DMSO
Content And Storage Store in freezer at -5°C to -30°C and protect from light.
Excitation Wavelength Range RNA: 499 nm, DNA: 500 nm
Dye Type SYTO 12 Green
For Use With (Application) Fluorescent Labeling, Live Cell Imaging, Microarray
For Use With (Equipment) Fluorescence Microscope, High Content Imager, Flow Cytometer
Storage Buffer DMSO
Quantity 315 μL
Volume (Metric) 315 μL
Detection Method Fluorescence
Emission RNA: 522 nm, DNA: 519 nm
Product Line SYTO
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Nucleic Acid Stain
Sub Cellular Localization Nucleic Acids
Show More Show Less
How do SYTO dyes bind to DNA?

The binding mode of SYTO nucleic acid stains is unknown. However, the behavior of these and related nucleic acid dyes suggests the following binding properties:

1.They appear to contact the solvent (suggested by sensitivity to salt, divalent cations, and in particular, SDS) and thus are likely to have contacts in the grooves.
2.All SYTO dyes appear to show some base selectivity and are thus likely to have minor groove contacts.
3.They can be removed from nucleic acid via ethanol precipitation; this characteristic is not shared by ethidium bromide and other intercalators. Likewise, the dyes are not removed from nucleic acid via butanol or chloroform extraction. These extraction methods do remove ethidium bromide from nucleic acid. 4. SYTO binding is not affected by nonionic detergents.
5. SYTO dyes are not quenched by BrdU, so they do not bind nucleic acids in precisely the same way as Hoechst 33342 and DAPI ((4′,6-diamidino-2-phenylindole).

SYBR Green I has shown little mutagenicity on frameshift indicator strains, indicating that it isn't likely to strongly intercalate.

What are the best nucleic acid stains for labeling nuclei in live cells in green and red?

SYTO 9 dye is one good choice for green; it is similar to fluorescein in excitation and emission wavelength. SYTO 59 dye is a good choice for red; it is similar to Texas Red. Both dyes are cell-permeant and stain DNA well, resulting in good nuclear labeling in live cells. With too high a concentration of dye and/or too long an incubation time, both may also label RNA, resulting in cytoplasmic and nucleolar staining, particularly with SYTO 59 dye.

I have a LIVE/DEAD BacLight Bacterial Viability kit that has SYTO 9 and propidium iodide in it. Will I be able to stain eukaryotic cells that have engulfed bacteria and determine if the bacteria are alive or dead using this kit?

Unfortunately, no. SYTO 9 will label the nuclei of live or dead cells, including the eukaryotic cells. Propidium iodide is cell impermeant, and will only enter dead cells. If the eukaryotic cells are dead, they will label with propidium iodide as well. If the eukaryotic cells are alive, propidium iodide will not be able to enter and thus will not label the bacteria inside, whether the bacteria are alive or dead. We are not aware of any way to do a viability assay of bacteria once they have been engulfed by cells.

I am having a hard time separating my bacteria from instrument noise. How can I optimize my signal?

Electronic noise is due to stray light collected by the system optics and/or low-level electronic signals. Noise may be eliminated or reduced through correct setup of instrument threshold and photomultiplier tube (PMT) voltage. The easiest way to differentiate a bacterial population from other small particles and electronic noise is to label the bacteria with a fluorescent stain and use the fluorescence emission to identify the population of interest. SYTO 9 is a great stain to label all cells and is detected in the FITC channel.

For Research Use Only. Not for use in diagnostic procedures.