missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYT12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5120539 Supplier Invitrogen™ Supplier No. PA5120539
Only null left

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain. Immunogen sequence: KGSLSIEDTF ESISELGPLE LMGRELDLAP YGTLRKSQSA DSLNSISSVS NTFGQDFTLG QVEVSMEYDT ASHTLNVAVM QGKDLLEREE ASFESCFMRV SLLPDE.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. Studies of the orthologous gene in rat have shown that the encoded protein selectively modulates spontaneous synaptic-vesicle exocytosis and may also be involved in regulating calcium independent secretion in nonneuronal cells. Alternative splicing results in multiple transcript variants. The gene has previously been referred to as synaptotagmin XI but has been renamed synaptotagmin XII to be standard with mouse and rat official nomenclature.

Specifications

Antigen SYT12
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SYT12
Gene Accession No. Q920N7
Gene Alias Srg1; synaptotagmin 12; synaptotagmin XII; Synaptotagmin12; Synaptotagmin-12; synaptotagmin-related gene 1 protein; SynaptotagminXII; synaptotagmin-XII; Syt XII; SYT11; Syt12; SYT-12; Sytr1; SytXII
Gene Symbols SYT12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 95-200 of human SYT12 (NP_001171351.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171180
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less