missing translation for 'onlineSavingsMsg'
Learn More

Syringic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014478
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014478 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014478 Supplier Merck Emd Millipore Supplier No. 63627100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 530-57-4
Physical Form Neat
Linear Formula HOC6H2(OCH3)2 CO2 H
MDL Number MFCD00002552
Melting Point 205°C to 209°C
Synonym 3,5-Dimethoxy-4-hydroxybenzoic acid; 4-Hydroxy-3,5-dimethoxy-benzoic acid; Gallic acid 3,5-dimethyl ether
Type Analytical Standard
Quantity 100 mg
Formula Weight 198.17
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less