missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Syntide 2, 96%

Catalog No. AAJ62843MCR
Change view
Click to view available options
Quantity:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62843MCR 1mg
AAJ62843LB0 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62843MCR Supplier Thermo Scientific Chemicals Supplier No. J62843MCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For research use

Selective substrate for calmodulin-dependent protein kinase II

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

For Use With (Application) Enzyme Assay Substrate
Formulation Lyophilized powder
Quantity 1mg
Storage Requirements -30° to -10°C
Regulatory Status RUO
Protein Length Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-gly-Leu-Pro-gly-Lys-Lys
Product Type Syntide 2, 96%
Recombinant Native
Form Lyophilized
Purity or Quality Grade 96%

RUO – Research Use Only