missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Syntide 2, 96%
For research use
Supplier: Thermo Scientific Chemicals J62843LB0
Description
Selective substrate for calmodulin-dependent protein kinase II
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| Enzyme Assay Substrate | |
| -30° to -10°C | |
| Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-gly-Leu-Pro-gly-Lys-Lys | |
| Native | |
| 96% |
| Lyophilized powder | |
| RUO | |
| Syntide 2, 96% | |
| Lyophilized |
Specifications
| Enzyme Assay Substrate | |
| 5mg | |
| RUO | |
| Syntide 2, 96% | |
| Lyophilized |
| Lyophilized powder | |
| -30° to -10°C | |
| Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-gly-Leu-Pro-gly-Lys-Lys | |
| Native | |
| 96% |
RUO – Research Use Only