missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Vigreux Distilling Columns

Indentated for improved vapor-liquid contact
Supplier: Synthware™ C101922
Description
- Column length measured from highest to lowest indent
- With top outer and lower inner standard taper joints
Chemical Identifiers
| Vigreux Column |
Specifications
| 120 mm | |
| Indentation: 120mm | |
| Vigreux Column | |
| Upper: 19/22; Lower: 19/22 |