missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Twin Connecting Hose Adapters
Two hose connections 180° from one another to permit a flow of inert gas over the apparatus
Supplier: Synthware™ A341922
Description
Synthware Inert Gas Adapter has two hose connections 180 degrees from one another to provide a flow of inert gas over the apparatus. The joints are standard taper inner joints.Specifications
| Twin Connecting Hose Adapters | |
| Hose O.D.: 8 mm | |
| Twin Connecting Hose Adapters | |
| 19/22 |
| Organic Synthesis | |
| Borosilicate Glass (3.3) | |
| 1 |