Learn More
Synthware™ Three Neck Flask, Vertical
Three Neck Flask with standard taper outer joints on both center and vertical side necks.
Supplier:  Synthware™ F44451L
 View more versions of this product
Catalog No. 31501285
Only null left
Explore available promotions
Description
Description
  • All sizes are hand blown from tubing to ensure uniform wall thickness.
Specifications
Specifications
Three Neck Round Bottom Flask, Vertical
Borosilicate Glass (3.3)
Center: 45/50; 24/40 side
Reusable
Beaded
Round
1 L
Three-neck Flask
Autoclavable
No
Non-sterile
Product Suggestions
Product Suggestions