Learn More
Synthware™ Three Neck Round Bottom Flasks, Vertical Side Neck with Bottom Outlet
Description
- All sizes are hand blown from tubing to ensure uniform wall thickness
- 4 mm straight bore PTFE stopcock lower outlet
Specifications
Specifications
| Description | Round Bottom Flask, 3-Neck Bottom Outlet |
| Capacity (Metric) | 2000 mL |
| Standard Taper Joint | Center: 45/50; 24/40 side |