missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Sublimator, with 2mm Glass Stopcock
GREENER_CHOICE

Catalog No. 31502087
Change view
Click to view available options
Standard Taper Joint:
24/40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31502087 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31502087 Supplier Synthware™ Supplier No. D232440G

With 2mm Glass Stopcock

  • Supplied with special 25mL long-neck flask for small volume efficiency

Specifications

Material Glass
Standard Taper Joint 24/40
Description 2mm Glass stopcock