Learn More
Synthware™ Snyder Distillation Column
GREENER_CHOICE
Catalog No. 31501085
Change view
Click to view available options
Standard Taper Joint:
24/40
Catalog No. Standard Taper Joint
31501085 24/40
1 options
Catalog No. 31501085 Supplier Synthware™ Supplier No. C302440
For use with Kuderna-Danish flask
  • With upper outer and lower inner standard taper joints
  • Both upper outer and lower inner standard taper joints of same size
Specifications
Height (Metric) 300mm
Type Snyder Distillation Column
Standard Taper Joint 24/40