Learn More
Synthware™ Separatory Funnel, with Glass Stopper
Separatory Funnel includes glass stopper.
Supplier: Synthware™ F471660A
Description
- With upper outer standard taper joint.
Spécifications
| 60 mL | |
| No.16 | |
| Glass |
| Borosilicate Glass (3.3) | |
| 2 mm | |
| Separatory Funnel |