missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Round Bottom Flasks, 4-Neck

Description
- All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
| Color | Clear |
| Description | Round bottom flask, 4-Neck |
| Capacity (Metric) | 5 L |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Organic Synthesis |
| Standard Taper Joint | Center: 45/50; Side: 24/40 |