Learn More
Synthware™ Recovery Flasks
Used as an evaporation flask in rotary evaporators
Supplier: Synthware™ F318500
Description
- WIth standard taper outer joint
- Heavy wall design for increased safety
- All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
| Recovery flask | |
| 500 mL | |
| 29/42 |