Learn More
Synthware™ Recovery Flasks
Used as an evaporation flask in rotary evaporators
Supplier:  Synthware™ F311410
 Afficher plus de versions de ce produit
Numéro de catalogue. 31501156
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • WIth standard taper outer joint
  • Heavy wall design for increased safety
  • All sizes are hand blown from tubing to ensure uniform wall thickness
Spécifications
Spécifications
Recovery flask
10 mL
14/20
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS